accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOIIHO

CAS RN 133-91-5

IUPAC Name: 2-hydroxy-3,5-diiodobenzoic acid
Synonyms: 3,5-diiodosalicylic acid
Molecular Weight (g/mol): 389.92
Molecular Formula: C7H4I2O3
InChi Key: DHZVWQPHNWDCFS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(I)=CC(I)=C1O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)