accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1330-38-7

IUPAC Name: copper(2+) disodium 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide-15,33-disulfonate
Synonyms: copper(2+) disodium 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide-15,33-disulfonate
Molecular Weight (g/mol): 780.16
Molecular Formula: C32H14CuN8Na2O6S2
InChi Key: LSASILAMYYWGBH-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[Cu++].[O-]S(=O)(=O)C1=CC=C2C3=NC4=NC(=NC5=C6C=C(C=CC6=C([N-]5)N=C5N=C(N=C([N-]3)C2=C1)C1=CC=CC=C51)S([O-])(=O)=O)C1=CC=CC=C41
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results