accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOS

CAS RN 133044-44-7

IUPAC Name: 6-acetyl-2,3-dihydro-1,3-benzothiazol-2-one
Synonyms: 6-acetyl-3H-1,3-benzothiazol-2-one
Molecular Weight (g/mol): 193.22
Molecular Formula: C9H7NO2S
InChi Key: UFRAIEFXNRTICG-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2NC(=O)SC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results