accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133057-83-7

IUPAC Name: [4-(benzyloxy)-3-fluorophenyl]boronic acid
Synonyms: 4-(benzyloxy)-3-fluorophenylboronic acid
Molecular Weight (g/mol): 246.04
Molecular Formula: C13H12BFO3
InChi Key: DQPDLDQJMUGVGI-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(OCC2=CC=CC=C2)C(F)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results