accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFBr

CAS RN 133059-44-6

IUPAC Name: 4-bromo-3-fluorobenzonitrile
Synonyms: 4-bromo-3-fluorobenzonitrile
Molecular Weight (g/mol): 200.01
Molecular Formula: C7H3BrFN
InChi Key: QBKXYSXQKRNVRQ-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results