accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCOCH3OCH3MgBr

CAS RN 133095-91-7

IUPAC Name: magnesium(2+) 3,4,5-trimethoxybenzen-1-ide bromide
Synonyms: magnesium(2+) 3,4,5-trimethoxybenzen-1-ide bromide
Molecular Weight (g/mol): 271.39
Molecular Formula: C9H11BrMgO3
InChi Key: RXSCVXDQDANNME-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].COC1=C[C-]=CC(OC)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results