accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CH3CNCH3H3CH3CH3CNCH3H3CH3CIII

CAS RN 13311-45-0

IUPAC Name: tris(tetrabutylazanium) triiodide
Synonyms: tris(tetrabutylammonium ion) triiodide
Molecular Weight (g/mol): 1108.13
Molecular Formula: C48H108I3N3
InChi Key: MZYZRSVLEXUIRP-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results