accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNH2OFFFHCl

CAS RN 133115-72-7

IUPAC Name: [4-(trifluoromethoxy)phenyl]hydrazine hydrochloride
Synonyms: [4-(trifluoromethoxy)phenyl]hydrazine hydrochloride
Molecular Weight (g/mol): 228.6
Molecular Formula: C7H8ClF3N2O
InChi Key: KQXZVSQCMVKMBK-UHFFFAOYSA-N
SMILES: Cl.NNC1=CC=C(OC(F)(F)F)C=C1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)