accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONHHNONH2OHO

CAS RN 133174-15-9

IUPAC Name: 5-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 5-(carbamoylamino)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 397.43
Molecular Formula: C21H23N3O5
InChi Key: NBMSMZSRTIOFOK-UHFFFAOYNA-N
SMILES: NC(=O)NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)