accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHHNNClNClNSOHOOOOSHOOO

CAS RN 13324-20-4

IUPAC Name: 1-amino-4-({3-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]-4-sulfophenyl}amino)-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid
Synonyms: procion blue MX-R
Molecular Weight (g/mol): 637.42
Molecular Formula: C23H14Cl2N6O8S2
InChi Key: RTLULCVBFCRQKI-UHFFFAOYSA-N
SMILES: NC1=C(C=C(NC2=CC=C(C(NC3=NC(Cl)=NC(Cl)=N3)=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)