accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13325-10-5

IUPAC Name: 4-hydroxybutan-1-aminium
Synonyms: 4-hydroxybutan-1-aminium
Molecular Weight (g/mol): 90.15
Molecular Formula: C4H12NO
InChi Key: BLFRQYKZFKYQLO-UHFFFAOYSA-O
SMILES: [NH3+]CCCCO
Molecular Weight (g/mol) 
  • (8)
  • (5)
Quantity 
  • (4)
  • (3)
  • (6)
Boiling Point 
  • (7)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (2)
  • (8)
  • (8)
  • (5)
  • (4)
  • (3)
  • (6)
  • (7)
  • (2)
  • (3)
  • (2)
  • (10)
  • (7)