accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133338-85-9

IUPAC Name: N,N-dicyclohexyl-2-(2,2-di{[(dicyclohexylcarbamoyl)methoxy]methyl}butoxy)acetamide
Synonyms: N,N-dicyclohexyl-2-(2,2-di{[(dicyclohexylcarbamoyl)methoxy]methyl}butoxy)acetamide
Molecular Weight (g/mol): 798.21
Molecular Formula: C48H83N3O6
InChi Key: QSTVDAZAPLNMPD-UHFFFAOYSA-N
SMILES: CCC(COCC(=O)N(C1CCCCC1)C1CCCCC1)(COCC(=O)N(C1CCCCC1)C1CCCCC1)COCC(=O)N(C1CCCCC1)C1CCCCC1
Grade 
  • (1)
Product Line 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results