CAS RN 1334509-86-2
CAS RN 1334509-86-2
IUPAC Name:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1-methyl-1H-indol-3-yl)propanoic acid
Synonyms:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(1-methylindol-3-yl)propanoic acid
Molecular Weight (g/mol):
440.5
Molecular Formula:
C27H24N2O4
InChi Key:
GDPXIUSEXJJUGT-DEOSSOPVSA-N
SMILES:
CN1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Trp Me -OH, Advanced ChemTech
CAS: 1334509-86-2 Molecular Formula: C27H24N2O4 Molecular Weight (g/mol): 440.50 MDL Number: MFCD02684512 InChI Key: GDPXIUSEXJJUGT-DEOSSOPVSA-N IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1-methyl-1H-indol-3-yl)propanoic acid SMILES: CN1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12