CAS RN 1335243-55-4
IUPAC Name:
2-chloro-3-iodo-1,4-dimethoxybenzene
Synonyms:
2-chloro-3-iodo-1,4-dimethoxybenzene
Molecular Weight (g/mol):
298.5
Molecular Formula:
C8H8ClIO2
InChi Key:
IMMMDYMSUZKGSS-UHFFFAOYSA-N
SMILES:
COC1=C(Cl)C(I)=C(OC)C=C1
1
–
1
of
1
results
1
Promotions Available
2-Chloro-3-iodo-1,4-dimethoxybenzene, 97%
2-Chloro-3-iodo-1,4-dimethoxybenzene, 97%
CAS: 1335243-55-4 Molecular Formula: C8H8ClIO2 Molecular Weight (g/mol): 298.504 MDL Number: MFCD22380712 InChI Key: IMMMDYMSUZKGSS-UHFFFAOYSA-N Synonym: 1-chloro-3,6-dimethoxy-2-iodobenzene PubChem CID: 98001136 IUPAC Name: 2-chloro-3-iodo-1,4-dimethoxybenzene SMILES: COC1=C(C(=C(C=C1)OC)I)Cl