CAS RN 133550-18-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ AG 538
A potent, cell-premeable, reversible, and competitive inhibitor of IGF-1 receptor kinase (IC50 = 400 nM).