accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133550-35-3

IUPAC Name: 2-cyano-3-(3,4-dihydroxyphenyl)-N-phenylprop-2-enamide
Synonyms: 2-cyano-3-(3,4-dihydroxyphenyl)-N-phenylprop-2-enamide
Molecular Weight (g/mol): 280.28
Molecular Formula: C16H12N2O3
InChi Key: HKHOVJYOELRGMV-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C(C=C(C#N)C(=O)NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results