accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOOHOHN(E/Z)

CAS RN 133550-37-5

IUPAC Name: 2-cyano-3-(3,4-dihydroxyphenyl)-N-(1-phenylethyl)prop-2-enamide
Synonyms: 2-cyano-3-(3,4-dihydroxyphenyl)-N-(1-phenylethyl)prop-2-enamide
Molecular Weight (g/mol): 308.34
Molecular Formula: C18H16N2O3
InChi Key: UMGQVUWXNOJOSJ-UHFFFAOYNA-N
SMILES: CC(NC(=O)C(=CC1=CC(O)=C(O)C=C1)C#N)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results