accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3SClOO

CAS RN 13360-57-1

IUPAC Name: N,N-dimethylsulfamoyl chloride
Synonyms: dimethylsulphamoyl-chloride
Molecular Weight (g/mol): 143.59
Molecular Formula: C2H6ClNO2S
InChi Key: JFCHSQDLLFJHOA-UHFFFAOYSA-N
SMILES: CN(C)S(Cl)(=O)=O
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)