accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OON

CAS RN 13361-31-4

IUPAC Name: 2-methylpropyl 2-cyanoacetate
Synonyms: 2-methylpropyl 2-cyanoacetate
Molecular Weight (g/mol): 141.17
Molecular Formula: C7H11NO2
InChi Key: HRGQEKKNLHJZGZ-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)CC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results