accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13362-30-6

IUPAC Name: 2-amino-4-(ethoxycarbonyl)pyridin-1-ium
Synonyms: 2-amino-4-(ethoxycarbonyl)pyridin-1-ium
Molecular Weight (g/mol): 167.19
Molecular Formula: C8H11N2O2
InChi Key: XVBZFXZNJAFCHL-UHFFFAOYSA-O
SMILES: CCOC(=O)C1=CC=[NH+]C(N)=C1
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)