accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13363-51-4

IUPAC Name: benzene-1,4-dicarbothioamide
Synonyms: benzene-1,4-dicarbothioamide
Molecular Weight (g/mol): 196.29
Molecular Formula: C8H8N2S2
InChi Key: USHPIZCRGQUHGN-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(C=C1)C(N)=S
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results