accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3NO

CAS RN 13368-42-8

IUPAC Name: 4-(trimethylsilyl)morpholine
Synonyms: 4-(trimethylsilyl)morpholine
Molecular Weight (g/mol): 159.3
Molecular Formula: C7H17NOSi
InChi Key: JJOWIQMPCCUIGA-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N1CCOCC1
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)