accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13372-81-1

IUPAC Name: (E)-N-[(2E)-3-phenylprop-2-en-1-ylidene]hydroxylamine
Synonyms: (E)-N-[(2E)-3-phenylprop-2-en-1-ylidene]hydroxylamine
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: RUQDOYIAKHIMAN-DAAQNPAKSA-N
SMILES: O\N=C\C=C\C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results