accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNOO

CAS RN 13380-67-1

IUPAC Name: 1-(4-bromophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-bromophenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 252.07
Molecular Formula: C10H6BrNO2
InChi Key: FECSFBYOMHWJQG-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1C(=O)C=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results