accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133812-16-5

IUPAC Name: 4-benzyl-3-[(2E)-but-2-enoyl]-1,3-oxazolidin-2-one
Synonyms: 4-benzyl-3-[(2E)-but-2-enoyl]-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 245.28
Molecular Formula: C14H15NO3
InChi Key: UTZAFVPPWUIPBH-QHHAFSJGNA-N
SMILES: C\C=C\C(=O)N1C(CC2=CC=CC=C2)COC1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results