accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH3COHCH3CH2OOSH3CCH3HNOCH3H3CNH2CH3HCl

CAS RN 133868-46-9

IUPAC Name: 4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[1-(2-amino-3-methylbutanamido)-2-methylpropan-2-yl]sulfanyl}acetate hydrochloride
Synonyms: 4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[1-(2-amino-3-methylbutanamido)-2-methylpropan-2-yl]sulfanyl}acetate hydrochloride
Molecular Weight (g/mol): 601.28
Molecular Formula: C31H53ClN2O5S
InChi Key: MFBPRQKHDIVLOJ-UHFFFAOYNA-N
SMILES: Cl.CC(C)C(N)C(=O)NCC(C)(C)SCC(=O)OC1CC(C)(C=C)C(O)C(C)C23CCC(=O)C2C1(C)C(C)CC3
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)