accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133902-66-6

HONNNCl

CAS RN 133902-66-6

IUPAC Name: [1-(4-chlorophenyl)-1H-1,2,3-triazol-4-yl]methanol
Synonyms: [1-(4-chlorophenyl)-1,2,3-triazol-4-yl]methanol
Molecular Weight (g/mol): 209.63
Molecular Formula: C9H8ClN3O
InChi Key: CAHIFLPAMJOAGI-UHFFFAOYSA-N
SMILES: OCC1=CN(N=N1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results