CAS RN 13391-72-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dimethyl(dimethylamino)vinylsilane, 97%, Thermo Scientific™
CAS: 13391-72-5 Molecular Formula: C6H15NSi MDL Number: MFCD01318143 Synonym: (Dimethylamino)dimethylvinylsilane; (Dimethylamino)vinyldimethylsilane