accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHO

CAS RN 134-11-2

IUPAC Name: 2-ethoxybenzoic acid
Synonyms: 2-ethoxybenzoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: XDZMPRGFOOFSBL-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1C(O)=O
Percent Purity 
  • (2)
  • (12)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (11)
Physical Form 
  • (10)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (5)
  • (1)
  • (5)
  • (2)
  • (6)
  • (2)
  • (12)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (11)
  • (10)
  • (3)
  • (4)