accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2

CAS RN 134-32-7

IUPAC Name: naphthalen-1-amine
Synonyms: α naphthylamine α-naftylamin α-naftalamin naphthalidam 1-aminonaftalen 1-naftylamin alfa-naftyloamina alfanaftilamina C.I. azoic diazo component 114 fast garnet base B
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: RUFPHBVGCFYCNW-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)