accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134-62-3

IUPAC Name: N,N-diethyl-3-methylbenzamide
Synonyms: DETA epa pesticide chemical code 080301 caswell no. 346 deta-20 flypel Deet amincene C 140 repudin-special metadelphene n,n diethyltoluamide
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: MMOXZBCLCQITDF-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=CC(C)=C1
Molecular Weight (g/mol) 
  • (6)
  • (6)
  • (1)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (6)
  • (6)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (3)
  • (5)
  • (1)