accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134-84-9

IUPAC Name: (4-methylphenyl)(phenyl)methanone
Synonyms: methyl benzophenone
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: WXPWZZHELZEVPO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Boiling Point 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)