accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134-85-0

IUPAC Name: (4-chlorophenyl)(phenyl)methanone
Synonyms: (4-chlorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 216.66
Molecular Formula: C13H9ClO
InChi Key: UGVRJVHOJNYEHR-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Boiling Point 
  • (4)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (4)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (7)
  • (4)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)