accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3OHNCH3H3C

CAS RN 134-91-8

IUPAC Name: bis[4-(diethylamino)phenyl]methanol
Synonyms: bis[4-(diethylamino)phenyl]methanol
Molecular Weight (g/mol): 326.48
Molecular Formula: C21H30N2O
InChi Key: WCIQBUVXZZYFJP-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)C(O)C1=CC=C(C=C1)N(CC)CC
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results