accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH3CHO

CAS RN 134-96-3

IUPAC Name: 4-hydroxy-3,5-dimethoxybenzaldehyde
Synonyms: syringaldehyde
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: KCDXJAYRVLXPFO-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC(OC)=C1O
Boiling Point 
  • (5)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (9)
Percent Purity 
  • (3)
  • (3)
  • (5)
Quantity 
  • (1)
  • (3)
  • (4)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (5)
  • (3)
  • (3)
  • (2)
  • (9)
  • (3)
  • (3)
  • (5)
  • (1)
  • (3)
  • (4)
  • (3)
  • (6)