CAS RN 1340-02-9
IUPAC Name:
tetrasodium 2-[(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-{ethyl[(3-sulfonatophenyl)methyl]iminiumyl}cyclohexa-2,5-dien-1-ylidene)methyl]-5-hydroxybenzene-1-sulfonate bis(2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate)
Synonyms:
tetrasodium 2-[(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-{ethyl[(3-sulfonatophenyl)methyl]iminio}cyclohexa-2,5-dien-1-ylidene)methyl]-5-hydroxybenzenesulfonate bis(2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate)
Molecular Weight (g/mol):
1391.41
Molecular Formula:
C67H62N8Na4O14S3
InChi Key:
CNJVTYXLOYTWPD-UHFFFAOYSA-J
SMILES:
[Na+].[Na+].[Na+].[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O.CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O.CCN(CC1=CC(=CC=C1)S([O-])(=O)=O)C1=CC=C(C=C1)C(=C1C=CC(C=C1)=[N+](CC)CC1=CC(=CC=C1)S([O-])(=O)=O)C1=C(C=C(O)C=C1)S([O-])(=O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Methyl Purple, 0.1% w/v aq. soln.
Transition interval: pH 4.8 (purple) to pH 5.4 (green) | CAS: 1340-02-9