accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN(S)(R)

CAS RN 134003-03-5

IUPAC Name: (1S,4R)-2-azabicyclo[2.2.1]heptan-3-one
Synonyms: (1S,4R)-2-azabicyclo[2.2.1]heptan-3-one
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: UIVLZOWDXYXITH-UHNVWZDZSA-N
SMILES: O=C1N[C@H]2CC[C@@H]1C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results