accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OOCH3CH3H3CNH2HCl

CAS RN 13404-22-3

IUPAC Name: tert-butyl (2S)-2-aminopropanoate hydrochloride
Synonyms: tert-butyl (2S)-2-aminopropanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: WIQIWPPQGWGVHD-JEDNCBNOSA-N
SMILES: Cl.C[C@H](N)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)