accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134168-97-1

IUPAC Name: 3-bromo-5-fluoroaniline
Synonyms: 3-bromo-5-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: NGZAVSDIXFIWHJ-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results