accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(E)

CAS RN 13419-69-7

IUPAC Name: (2E)-hex-2-enoate
Synonyms: (2E)-hex-2-enoate
Molecular Weight (g/mol): 113.14
Molecular Formula: C6H9O2
InChi Key: NIONDZDPPYHYKY-SNAWJCMRSA-M
SMILES: CCC\C=C\C([O-])=O
Boiling Point 
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)