accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13421-13-1

IUPAC Name: 4-chloro-2-iodobenzoic acid
Synonyms: 4-chloro-2-iodobenzoic acid
Molecular Weight (g/mol): 282.46
Molecular Formula: C7H4ClIO2
InChi Key: LRRDANNSUCQNDU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C=C1I
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results