accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFN

CAS RN 134227-45-5

IUPAC Name: 3,4,5-trifluorobenzonitrile
Synonyms: 3,4,5-trifluorobenzonitrile
Molecular Weight (g/mol): 157.1
Molecular Formula: C7H2F3N
InChi Key: XFKYJMGXZXJYBS-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1F)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results