accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 13423-15-9

IUPAC Name: 3-methyloxolane
Synonyms: furan, tetrahydro-3-methyl-
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: LJPCNSSTRWGCMZ-UHFFFAOYNA-N
SMILES: CC1CCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results