accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH3CP

CAS RN 13423-48-8

IUPAC Name: heptyltriphenylphosphanium bromide
Synonyms: heptyltriphenylphosphanium bromide
Molecular Weight (g/mol): 441.39
Molecular Formula: C25H30BrP
InChi Key: WCZSOHSGMBVYFW-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results