accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13425-93-9

IUPAC Name: 6,7-dimethoxy-1,4-dihydroquinolin-4-one
Synonyms: 6,7-dimethoxy-1H-quinolin-4-one
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: QOGPNCUTXVZQSL-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(=O)C=CNC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results