accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHSNHNH2

CAS RN 13431-34-0

IUPAC Name: 3-amino-1-ethylthiourea
Synonyms: 3-amino-1-ethylthiourea
Molecular Weight (g/mol): 119.19
Molecular Formula: C3H9N3S
InChi Key: SKYYTGUCWARUCL-UHFFFAOYSA-N
SMILES: CCNC(=S)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results