accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134319-22-5

IUPAC Name: propyl 3-propoxypyridine-2-carboxylate
Synonyms: propyl 3-propoxypyridine-2-carboxylate
Molecular Weight (g/mol): 223.27
Molecular Formula: C12H17NO3
InChi Key: ZGUCGLNLZXLQTD-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=C(OCCC)C=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results