accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13433-31-3

IUPAC Name: 8-methoxy-1',3',3'-trimethyl-1',3'-dihydrospiro[chromene-2,2'-indole]
Synonyms: 8-methoxy-1',3',3'-trimethylspiro[chromene-2,2'-indole]
Molecular Weight (g/mol): 307.39
Molecular Formula: C20H21NO2
InChi Key: NQVQBIMDDKDYAO-UHFFFAOYNA-N
SMILES: COC1=CC=CC2=C1OC1(C=C2)N(C)C2=CC=CC=C2C1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results