accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13435-97-7

IUPAC Name: 2-[(2-phenylethenyl)sulfanyl]acetic acid
Synonyms: [(2-phenylethenyl)sulfanyl]acetic acid
Molecular Weight (g/mol): 194.25
Molecular Formula: C10H10O2S
InChi Key: IUEVBKDVPSQVLM-UHFFFAOYSA-N
SMILES: OC(=O)CSC=CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results