accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2ONH2

CAS RN 13438-65-8

IUPAC Name: 2-aminopyridine-3-carboxamide
Synonyms: 2-aminopyridine-3-carboxamide
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: HTPCDVLWYUXWQR-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)N=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results